missing translation for 'onlineSavingsMsg'
Learn More

3-Piperidinemethanol, 98%, ACROS Organics™

Brand:  Acros Organics 168040250

Code : 29

Additional Details : CAS Number : 4606-65-9 Weight : 0.07500kg Transport : UN number : 3263 Chem class : 1b Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11916311

  • £83.40 / 25g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3-Piperidinemethanol
97.5
98%
C6H13NO
3-(Hydroxymethyl)piperidine
4606-65-9
100.0
98%
115.17

Specifications

3-Piperidinemethanol
>112°C
Clear
25g
97.5
Authentic
1.02
98%
C6H13NO
3-(Hydroxymethyl)piperidine
1.0200g/mL
236.0°C
58.0° to 62.0°C
4606-65-9
100.0
Glass bottle
Liquid
98%
115.17
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Piperidinemethanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS