Learn More
Thermo Scientific Maybridge 3-Piperidinecarboxylic acid, 50g, 97%
Brand: Thermo Scientific Maybridge AC19919EE
3-Piperidinecarboxylic acid, 50g, 97%
Code : 31
Additional Details : CAS Number : 498-95-3 Weight : 0.10000kg
Brand: Thermo Scientific Maybridge AC19919EE
3-Piperidinecarboxylic acid, 50g, 97%
Code : 31
Additional Details : CAS Number : 498-95-3 Weight : 0.10000kg