missing translation for 'onlineSavingsMsg'
Learn More

3-Picoline-N-oxide, 98%, Thermo Scientific™

Catalog Number 10511381
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1003-73-2
C6H7NO
109.13
MFCD00006203
DMGGLIWGZFZLIY-UHFFFAOYSA-N
3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
13858
3-methyl-1-oxidopyridin-1-ium
CC1=C[N+](=CC=C1)[O-]
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1003-73-2
C6H7NO
109.13
MFCD00006203
DMGGLIWGZFZLIY-UHFFFAOYSA-N
3-methylpyridine 1-oxide, 3-picoline-n-oxide, 3-picoline n-oxide, 3-methylpyridine n-oxide, 3-methylpyridine-n-oxide, pyridine, 3-methyl-, 1-oxide, 3-picoline 1-oxide, beta-picoline n-oxide, 3-methylpyridin-1-ium-1-olate, 3-methylpyridine-1-oxide
13858
3-methyl-1-oxidopyridin-1-ium
CC1=C[N+](=CC=C1)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 40°C
Color Brown to Yellow
Boiling Point 150°C (15.0 mmHg)
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 500 g
Solubility Information Solubility in water: freely soluble in water
Formula Weight 109.13
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3-Picoline-N-oxide, 98%
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 3-Picoline-N-oxide
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'