missing translation for 'onlineSavingsMsg'
Learn More

3-Phthalimidopropionic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7058397
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
3339-73-9
C11H9NO4
219.196
MFCD00023096
DXXHRZUOTPMGEH-UHFFFAOYSA-N
3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
76859
3-(1,3-dioxoisoindol-2-yl)propanoic acid
C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O
This item is not returnable. View return policy
3339-73-9
C11H9NO4
219.196
MFCD00023096
DXXHRZUOTPMGEH-UHFFFAOYSA-N
3-phthalimidopropionic acid, 3-1,3-dioxoisoindolin-2-yl propanoic acid, 2h-isoindole-2-propanoic acid, 1,3-dihydro-1,3-dioxo, 3-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 3-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, pht-beta-ala-oh, b-phthalimidopropionic acid, 3-phthalimidopropanoic acid, n-phthaloyl-beta-alanine, phthalyl-.beta.-alanine
76859
3-(1,3-dioxoisoindol-2-yl)propanoic acid
C1=CC=C2C(=C1)C(=O)N(C2=O)CCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 151°C to 153°C
Quantity 1 g
Beilstein 15570
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material 3-Phthalimidopropionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'