missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylpropyl isocyanate, 97%, Thermo Scientific™

Catalog Number p-7077672
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
68664-23-3
C10H11NO
161.21
WPABAKMRZCBXTO-UHFFFAOYSA-N
3-phenylpropyl isocyanate, 3-phenylpropylisocyanate, 3-isocyanatopropyl benzene, phenylpropylisocyanate, acmc-20amyq, 3-isocyanatopropyl-benzene, 1-3-isocyanatopropyl benzene, 3-isocyanato-propyl-benzene, benzene, 3-isocyanatopropyl
3427666
3-isocyanatopropylbenzene
C1=CC=C(C=C1)CCCN=C=O
This item is not returnable. View return policy
68664-23-3
C10H11NO
161.21
WPABAKMRZCBXTO-UHFFFAOYSA-N
3-phenylpropyl isocyanate, 3-phenylpropylisocyanate, 3-isocyanatopropyl benzene, phenylpropylisocyanate, acmc-20amyq, 3-isocyanatopropyl-benzene, 1-3-isocyanatopropyl benzene, 3-isocyanato-propyl-benzene, benzene, 3-isocyanatopropyl
3427666
3-isocyanatopropylbenzene
C1=CC=C(C=C1)CCCN=C=O
Boiling Point 115.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 161.21
Percent Purity 97%
Chemical Name or Material 3-Phenylpropyl isocyanate
compliance-icons compliance-icons
Product Identifier
  • 3-Phenylpropyl isocyanate, 97%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'