missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpropiophenone, 98%, Thermo Scientific Chemicals
48.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
3-Phenylpropiophenone is used to produce 2-bromo-1,3-diphenyl-propan-1-one.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 68°C to 73°C |
Quantity | 10 g |
Beilstein | 1910661 |
Solubility Information | Insoluble in water. |
Formula Weight | 210.28 |
Percent Purity | 98% |
Chemical Name or Material | 3-Phenylpropiophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'