missing translation for 'onlineSavingsMsg'
Learn More

3-Phenylacrylaldehyde oxime, 97%, Maybridge™

£7.27 - £61.30

Chemical Identifiers

CAS 13372-81-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
InChI Key RUQDOYIAKHIMAN-DAAQNPAKSA-N
Synonym cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z
PubChem CID 9561350
IUPAC Name (NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine
SMILES C1=CC=C(C=C1)C=CC=NO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10649404 SDS View Documents Maybridge
BTB09548DA
1g £7.27
1g View alternative products
Save Up to 
»
Estimated Shipment
13-08-2020
Please to purchase this item. Need a web account? Register with us today!
10007102 SDS View Documents Maybridge
BTB09548EA
10g £28.05
10g View alternative products
Save Up to 
»
Estimated Shipment
13-07-2020
Please to purchase this item. Need a web account? Register with us today!
10624375 SDS View Documents Maybridge
BTB09548EB
25g £61.30
25g View alternative products
Save Up to 
»
Estimated Shipment
13-08-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

13372-81-1
147.177
cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z
(NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine
C9H9NO
RUQDOYIAKHIMAN-DAAQNPAKSA-N
9561350
C1=CC=C(C=C1)C=CC=NO

Specifications

3-Phenylacrylaldehyde oxime
13372-81-1
C9H9NO
RUQDOYIAKHIMAN-DAAQNPAKSA-N
(NE)-N-[(E)-3-phenylprop-2-enylidene]hydroxylamine
9561350
1g
97%
cinnamaldehyde oxime, 3-phenylacrylaldehyde oxime, phenylacrylaldehyde oxime, e-n-2e-3-phenylprop-2-en-1-ylidene hydroxylamine, n-hydroxy-3-phenylprop-2-en-1-imine, n-3-phenylprop-2-en-1-ylidene hydroxylamine, 3-phenylpropenal oxime, benzeneacrylaldehydeoxime, 3-phenyl-2-propenal oxime, cinnamaldoxime, e + z
C1=CC=C(C=C1)C=CC=NO
147.177
147.18