missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenyl-5-piperazino-1,2,4-thiadiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-phenyl-5-piperazino-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl piperazine, 3-phenyl-5-piperazin-1-yl-1,2,4-thiadiazole, piperazine, 1-3-phenyl-1,2,4-thiadiazol-5-yl, maybridge1_003948, 3-phenyl-5-piperazinyl-1,2,4-thiadiazole, 1-3-phenyl-1,2,4-thiadiazol-5-yl piperazin, 1-3-phenyl-1,2,4-thiadiazol-5-yl-piperazine, 1-3-phenyl-1,2,4 thiadiazol-5-yl-piperazine, piperazine,1-3-phenyl-1,2,4-thiadiazol-5-yl |
Formula Weight | 246.34 |
Percent Purity | 97% |
Chemical Name or Material | 3-Phenyl-5-piperazino-1, 2, 4-thiadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'