missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63692.03

Code : NEW

Additional Details : CAS Number : 2010-15-3 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15450519

  • £109.18 / 1g


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Phenyl-2-thiohydantoin
192.24
UN2811
2010-15-3
MFCD00027413
3-Phenyl-2-thioxo-4-imidazolidinone

Specifications

3-Phenyl-2-thiohydantoin
1g
Solid
192.24
UN2811
Yellow
2010-15-3
95%
MFCD00027413
3-Phenyl-2-thioxo-4-imidazolidinone
Videos
SDS