missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2-propyn-1-ol, 98+%, Thermo Scientific Chemicals

Catalog Number p-7059019
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
1504-58-1
C9H8O
132.162
MFCD00040914
NITUNGCLDSFVDL-UHFFFAOYSA-N
3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
123115
3-phenylprop-2-yn-1-ol
C1=CC=C(C=C1)C#CCO
This item is not returnable. View return policy
1504-58-1
C9H8O
132.162
MFCD00040914
NITUNGCLDSFVDL-UHFFFAOYSA-N
3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
123115
3-phenylprop-2-yn-1-ol
C1=CC=C(C=C1)C#CCO

3-Phenyl-2-propyn-1-ol is an phenyl acetylene derivative with inhibitory effect on nitrification in soil.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.078 g/mL
Boiling Point 129°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5867
Quantity 2 g
Beilstein 2040911
Solubility Information Soluble in ether,acetone,benzene,chloroform,ethyl Acetate,methanol.
Formula Weight 132.16
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-2-propyn-1-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'