missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-1-propyne, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3779449
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10147-11-2
C9H8
116.16
MFCD00134431
NGKSKVYWPINGLI-UHFFFAOYSA-N
3-phenyl-1-propyne, 2-propynyl benzene, benzylacetylene, prop-2-yn-1-ylbenzene, benzene, 2-propynyl, propargylbenzol, propargylbenzene, benzyl acetylene, 3-phenyl-propyne, 3-phenyl-1-propyn
575753
prop-2-ynylbenzene
C#CCC1=CC=CC=C1
This item is not returnable. View return policy
10147-11-2
C9H8
116.16
MFCD00134431
NGKSKVYWPINGLI-UHFFFAOYSA-N
3-phenyl-1-propyne, 2-propynyl benzene, benzylacetylene, prop-2-yn-1-ylbenzene, benzene, 2-propynyl, propargylbenzol, propargylbenzene, benzyl acetylene, 3-phenyl-propyne, 3-phenyl-1-propyn
575753
prop-2-ynylbenzene
C#CCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9340g/mL
Boiling Point 75.0°C (20.0 mmHg)
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5260 to 1.5290
Quantity 1 g
Specific Gravity 0.934
Stabilizer 250ppm BHT
Formula Weight 116.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Phenyl-1-propyne, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Phenyl-1-propyne, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'