Learn More
Thermo Scientific Maybridge 3-(Phenoxymethyl)benzylamine, 1g, 97%
Brand: Thermo Scientific Maybridge CC63613DA
3-(Phenoxymethyl)benzylamine, 1g, 97%
Code : 31
Additional Details : CAS Number : 871893-47-9 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III