missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog Number p-180623
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
13826-35-2
C13H12O2
200.24
MFCD00004636
KGANAERDZBAECK-UHFFFAOYSA-N
3-phenoxybenzyl alcohol, 3-phenoxyphenyl methanol, 3-phenoxybenzylalcohol, m-phenoxybenzyl alcohol, 3-phenoxybenzenemethanol, benzenemethanol, 3-phenoxy, 3-hydroxymethyl diphenyl ether, 3-phenoxybenzylic alcohol, benzyl alcohol, m-phenoxy, 3-pboh
26295
CHEBI:62527
(3-phenoxyphenyl)methanol
C1=CC=C(C=C1)OC2=CC=CC(=C2)CO
This item is not returnable. View return policy
13826-35-2
C13H12O2
200.24
MFCD00004636
KGANAERDZBAECK-UHFFFAOYSA-N
3-phenoxybenzyl alcohol, 3-phenoxyphenyl methanol, 3-phenoxybenzylalcohol, m-phenoxybenzyl alcohol, 3-phenoxybenzenemethanol, benzenemethanol, 3-phenoxy, 3-hydroxymethyl diphenyl ether, 3-phenoxybenzylic alcohol, benzyl alcohol, m-phenoxy, 3-pboh
26295
CHEBI:62527
(3-phenoxyphenyl)methanol
C1=CC=C(C=C1)OC2=CC=CC(=C2)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless or White
Density 1.1456g/mL
Boiling Point 135.0°C to 140.0°C (1.0 mmHg)
Flash Point 309°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula C6H5OC6H4CH2OH
Refractive Index 1.5920 to 1.5940
Quantity 5 g
Specific Gravity 1.1456
Solubility Information Solubility in water: insoluble in water.
Formula Weight 200.24
Percent Purity 98%
Physical Form Liquid or Solid
Chemical Name or Material 3-Phenoxybenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Phenoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'