missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenoxybenzeneboronic acid, 97+%, Thermo Scientific Chemicals
35.46 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 138°C to 143°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble. |
Formula Weight | 214.03 |
Percent Purity | ≥97% |
Chemical Name or Material | 3-Phenoxybenzeneboronic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'