missing translation for 'onlineSavingsMsg'
Learn More

3-Penten-2-one, 70%, technical, Thermo Scientific Chemicals

Catalog Number p-4228861
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
625-33-2
C5H8O
84.12
MFCD00009290
LABTWGUMFABVFG-ONEGZZNKSA-N
z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
637920
(E)-pent-3-en-2-one
CC=CC(=O)C
This item is not returnable. View return policy
625-33-2
C5H8O
84.12
MFCD00009290
LABTWGUMFABVFG-ONEGZZNKSA-N
z-pent-3-en-2-one, unii-w8xx21whyc, z-3-penten-2-one, w8xx21whyc, 3-penten-2-one, 3z-pent-3-en-2-one, 3-penten-2-one, z, cis-3-penten-2-one, 3-penten-2-one, cis
637920
(E)-pent-3-en-2-one
CC=CC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 65.0
CAS Max % 100.0
Color Colorless
Density 0.8600g/mL
Boiling Point 121.0°C to 124.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 70% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=CHCOCH3
Refractive Index 1.4405
Quantity 1 g
Beilstein 01,732
Fieser 02,306; 03,218; 04,371
Specific Gravity 0.86
Formula Weight 84.12
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-Penten-2-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Penten-2-one, 70%, Technical Grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'