missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3'-(Pentafluorothio)acetophenone, 97%

Brand:  Alfa Aesar™ H33532.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426357

  • £60.60 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3'-(Pentafluorothio)acetophenone
246.19
MFCD16652407
3'-(Pentafluorosulfur)acetophenone

Specifications

3'-(Pentafluorothio)acetophenone
43° to 45°C
Crystals
246.19
3'-(Pentafluorosulfur)acetophenone
White
1g
97%
MFCD16652407
Videos
SDS