missing translation for 'onlineSavingsMsg'
Learn More

3-(p-Tolyl)propionic acid, 97%, Thermo Scientific™

Catalog Number 10483264
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1505-50-6
C10H12O2
164.2
MFCD00016561
LDYGRLNSOKABMM-UHFFFAOYSA-N
3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
73927
3-(4-methylphenyl)propanoic acid
CC1=CC=C(CCC(O)=O)C=C1
This item is not returnable. View return policy
1505-50-6
C10H12O2
164.2
MFCD00016561
LDYGRLNSOKABMM-UHFFFAOYSA-N
3-4-methylphenyl propionic acid, 3-p-tolyl propionic acid, 3-p-tolyl propanoic acid, benzenepropanoic acid, 4-methyl, 3-4-methylphenyl propanoic acid, 4-methylphenylpropionic acid, 3-p-tolylpropanoic acid, p-methylhydrocinnamic acid, 4-methyl phenyl propionic acid, 4-methylhydrocinnamic acid
73927
3-(4-methylphenyl)propanoic acid
CC1=CC=C(CCC(O)=O)C=C1
CAS Min % 96
CAS Max % 100
Melting Point 115.0°C to 118.0°C
Color Orange or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula HOOC(CH2)2C6H4CH3
Quantity 5 g
Formula Weight 164.2
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-(p-Tolyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-(p-Tolyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'