3-Oxopentanoic acid, 85%, Acros Organics


Overview
Brand: Acros Organics

Manufacturer Part Number: 452760050

UNSPSC: 12000000

Code: NEW

Additional Details:
Additional Details: CAS Number: 10191-25-0 Weight: 0.01000kg


Quantity 5g

Product Code. 15249836

Quantity Price
1 £ 269.9 / 5g
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-Oxopentanoic acid
CAS 10191-25-0
Purity Grade Technical Grade
Assay Percent Range 85%
Molecular Formula C5H8O3
Formula Weight 116.12

Specification

Formula Weight 116.12
Purity Grade Technical Grade
Quantity 5g
CAS 10191-25-0
Assay Percent Range 85%
Molecular Formula C5H8O3
Chemical Name or Material 3-Oxopentanoic acid