missing translation for 'onlineSavingsMsg'
Learn More

3-Oxobutanenitrile, 96%, Thermo Scientific Chemicals

Catalog Number 15471799
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2469-99-0
C4H5NO
83.09
MFCD05861381
OPXYNEYEDHAXOM-UHFFFAOYSA-N
3-oxobutyronitrile, 3-oxo-butyronitrile, butanenitrile, 3-oxo, cyanoacetone, cyanaceton, a-cyanoacetone, acetoacetonitrile, acetylacetonitrile, 1-cyano-2-propanone, alpha-acetylacetonitrile
75579
3-oxobutanenitrile
CC(=O)CC#N
This item is not returnable. View return policy
2469-99-0
C4H5NO
83.09
MFCD05861381
OPXYNEYEDHAXOM-UHFFFAOYSA-N
3-oxobutyronitrile, 3-oxo-butyronitrile, butanenitrile, 3-oxo, cyanoacetone, cyanaceton, a-cyanoacetone, acetoacetonitrile, acetylacetonitrile, 1-cyano-2-propanone, alpha-acetylacetonitrile
75579
3-oxobutanenitrile
CC(=O)CC#N

It is used as pharmaceutical raw material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Boiling Point 92°C to 94°C (10 mmHg)
Linear Formula CH3COCH2CN
Refractive Index 1.4285
Quantity 1 g
UN Number UN3276
Beilstein 1633584
Solubility Information Slightly soluble in water.
Formula Weight 83.09
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Oxobutanenitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'