missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CF3OC6H4C(O)CH2CN |
Quantity | 1 g |
Formula Weight | 229.16 |
Percent Purity | 97% |
Chemical Name or Material | 3-Oxo-3-[4-(trifluoromethoxy)phenyl]propanenitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'