missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Oxabicyclo[3.1.0]hexane-2,4-dione, ≥97%

Brand:  Alfa Aesar™ H64382.03

Code : NEW

Additional Details : CAS Number : 5617-74-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15421569

  • £53.70 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Oxabicyclo[3.1.0]hexane-2,4-dione
112.08
113493
5617-74-3
MFCD00126929
1,2-Cyclopropanedicarboxylic anhydride

Specifications

3-Oxabicyclo[3.1.0]hexane-2,4-dione
1.50g/mL
≥97%
MFCD00126929
1,2-Cyclopropanedicarboxylic anhydride
White
1g
5617-74-3
Powder
112.08
113493
100° to 102°C (5mmHg)
59° to 61°C (lit.)
Moisture Sensitive
Videos
SDS