missing translation for 'onlineSavingsMsg'
Learn More

3-Octylthiophene, 97%, Thermo Scientific Chemicals

Catalog Number p-176792
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
65016-62-8
MFCD00085281
WQYWXQCOYRZFAV-UHFFFAOYSA-N
3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
566852
3-octylthiophene
CCCCCCCCC1=CSC=C1
This item is not returnable. View return policy
65016-62-8
MFCD00085281
WQYWXQCOYRZFAV-UHFFFAOYSA-N
3-n-octylthiophene, thiophene, 3-octyl, acmc-1bfcd, 3-octylthiophene, ksc491i4b, bidd:gt0691
566852
3-octylthiophene
CCCCCCCCC1=CSC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 92.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 106°C to 107°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C12H20S
Refractive Index 1.4870 to 1.492
Quantity 10 g
Specific Gravity 0.91
Molecular Weight (g/mol) 196.36
Formula Weight 196.36
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Octylthiophene, 94%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Octylthiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'