missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Octanone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
3-Octanone was used to study the effect of TiO2 photo catalyst on the rate and the ratio of products generated in photo catalytic oxidation of 3-octanone. It is used as a flavor and fragrance ingredient.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Incompatible with strong oxidizing agents, strong reducing agents, strong bases.
Specifications
Melting Point | -23°C |
Boiling Point | 167°C to 168°C |
Flash Point | 51°C (123°F) |
Quantity | 25 g |
UN Number | UN1224 |
Beilstein | 1700021 |
Merck Index | 14,6084 |
Formula Weight | 128.22 |
Percent Purity | 98% |
Chemical Name or Material | 3-Octanone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'