missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrophenylsulfur pentafluoride, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It has a wide range of applications as a fluorinating agent. g. It is a convenient alternative to current fluorinating agents for use in the synthesis of specialty chemicals and intermediates, such as pharmaceuticals, agrochemicals
It has a wide range of applications as a fluorinating agent. g. It is a convenient alternative to current fluorinating agents for use in the synthesis of specialty chemicals and intermediates, such as pharmaceuticals, agrochemicals
Specifications
CAS | 2613-26-5 |
Density | 1.662 |
Boiling Point | 106°C (2 mmHg) |
Molecular Formula | C6H4F5NO2S |
Refractive Index | 1.476 |
MDL Number | MFCD00221621 |
Quantity | 1 g |
UN Number | UN2810 |
Beilstein | 2699206 |
Synonym | 3-nitrophenylsulfur pentafluoride, 3-nitrophenylsulphur pentafluoride, 1-nitro-3-pentafluorosulfanyl benzene, 1-nitro-3-pentafluoro-??-sulfanyl benzene, acmc-209goj, pentafluoro-3-nitrophenyl, 3-pentafluorothio nitrobenzene, 3-nitropentafluorosulfanylbenzene, 3-pentafluorosulphanylnitrobenzene, 3-pentafluorosulfur-nitrobenzene |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'