missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylacetic acid, 99%, ACROS Organics™

Brand:  Acros Organics 147050250

Code : 29

Additional Details : CAS Number : 1877-73-2 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10627124

  • £622.20 / 25g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3-Nitrophenylacetic acid
98.5
99%
C8H7NO4
181.15
1877-73-2
100.0
99%
O2NC6H4CH2CO2H
MFCD00007278

Specifications

3-Nitrophenylacetic acid
116.0° to 120.0°C
1877-73-2
100.0
Glass bottle
99%
C8H7NO4
181.15
09, 455
Cream
25g
98.5
Authentic
Crystalline Powder
99%
O2NC6H4CH2CO2H
MFCD00007278
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Nitrophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS