missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylacetic acid, 99%, Thermo Scientific™

Catalog Number p-3779310
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1877-73-2
MFCD00007278
WUKHOVCMWXMOOA-UHFFFAOYSA-N
3-nitrophenylacetic acid, 2-3-nitrophenyl acetic acid, 3-nitrophenyl acetic acid, benzeneacetic acid, 3-nitro, m-nitrophenyl acetic acid, m-nitrophenylacetic acid, 3-nitrobenzeneacetic acid, acetic acid, m-nitrophenyl, ccris 2337
15876
C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O
This item is not returnable. View return policy
1877-73-2
MFCD00007278
WUKHOVCMWXMOOA-UHFFFAOYSA-N
3-nitrophenylacetic acid, 2-3-nitrophenyl acetic acid, 3-nitrophenyl acetic acid, benzeneacetic acid, 3-nitro, m-nitrophenyl acetic acid, m-nitrophenylacetic acid, 3-nitrobenzeneacetic acid, acetic acid, m-nitrophenyl, ccris 2337
15876
C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 116.0°C to 120.0°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H7NO4
Linear Formula O2NC6H4CH2CO2H
Quantity 1 g
Beilstein 09, 455
Molecular Weight (g/mol) 181.147
Formula Weight 181.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'