missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrophenylacetic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Nitrophenylacetic acid, is a useful synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Nitrophenylacetic acid, is a useful synthetic intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
3-Nitrophenylacetic acid, is a useful synthetic intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
Specifications
Melting Point | 117°C to 120°C |
Quantity | 1 g |
Beilstein | 2050088 |
Solubility Information | Soluble in water. |
Formula Weight | 181.15 |
Percent Purity | 99% |
Chemical Name or Material | 3-Nitrophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'