missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog Number p-3758086
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3958-57-4
C7H6BrNO2
216.03
MFCD00007271
LNWXALCHPJANMJ-UHFFFAOYSA-N
3-nitrobenzyl bromide, 1-bromomethyl-3-nitrobenzene, m-nitrobenzyl bromide, benzene, 1-bromomethyl-3-nitro, alpha-bromo-3-nitrotoluene, m-bromomethyl nitrobenzene, alpha-bromo-m-nitrotoluene, m-nitro-alpha-bromotoluene, 3-nitrobenzylbromide, toluene, alpha-bromo-m-nitro
77568
1-(bromomethyl)-3-nitrobenzene
C1=CC(=CC(=C1)[N+](=O)[O-])CBr
This item is not returnable. View return policy
3958-57-4
C7H6BrNO2
216.03
MFCD00007271
LNWXALCHPJANMJ-UHFFFAOYSA-N
3-nitrobenzyl bromide, 1-bromomethyl-3-nitrobenzene, m-nitrobenzyl bromide, benzene, 1-bromomethyl-3-nitro, alpha-bromo-3-nitrotoluene, m-bromomethyl nitrobenzene, alpha-bromo-m-nitrotoluene, m-nitro-alpha-bromotoluene, 3-nitrobenzylbromide, toluene, alpha-bromo-m-nitro
77568
1-(bromomethyl)-3-nitrobenzene
C1=CC(=CC(=C1)[N+](=O)[O-])CBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 54°C to 58°C
Color Yellow to Brown
Boiling Point 153°C to 154°C (7.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2Br
Quantity 5 g
Beilstein 05, 334
Solubility Information Solubility in water: insoluble
Formula Weight 216.03
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Nitrobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3-Nitrobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'