missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-172205
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
121-51-7
C6H4ClNO4S
221.62
MFCD00007435
MWWNNNAOGWPTQY-UHFFFAOYSA-N
3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u
8476
3-nitrobenzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
121-51-7
C6H4ClNO4S
221.62
MFCD00007435
MWWNNNAOGWPTQY-UHFFFAOYSA-N
3-nitrophenylsulfonyl chloride, 3-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 3-nitro, 3-nitrobenzenesulphonyl chloride, 3-nitro-benzenesulfonyl chloride, m-nitrophenylsulfonyl chloride, m-nitrobenzenesulfonyl chloride, unii-kn1z8wpl9u, benzenesulfonyl chloride, m-nitro, kn1z8wpl9u
8476
3-nitrobenzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 60.0°C to 65.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula SO2ClC6H4NO2
Quantity 25 g
Solubility Information Solubility in water: decomposes
Formula Weight 221.62
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonyl chloride, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'