missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonic acid, sodium salt, 99%, Thermo Scientific Chemicals

Catalog Number p-3737057
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
127-68-4
C6H4NNaO5S
225.15
MFCD00007490
LJRGBERXYNQPJI-UHFFFAOYSA-M
sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
31389
sodium;3-nitrobenzenesulfonate
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]
127-68-4
C6H4NNaO5S
225.15
MFCD00007490
LJRGBERXYNQPJI-UHFFFAOYSA-M
sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6
31389
sodium;3-nitrobenzenesulfonate
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 350.0°C
Color Yellow
pH 6 to 10 (1% aq. soln.)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic Bottle
Linear Formula O2NC6H4SO3Na
Quantity 1 kg
Beilstein 11, 68
Solubility Information Solubility in water: 200g/L water (20°C)
Formula Weight 225.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonic acid, sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'