missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Nitrobenzenesulfonamide, 99%

Brand:  Alfa Aesar™ H55831.03

Code : NEW

Additional Details : CAS Number : 121-52-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15422198

  • £21.00 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Nitrobenzenesulfonamide
121-52-8
202.19
MFCD00007935

Specifications

3-Nitrobenzenesulfonamide
166° to 168°C
121-52-8
99%
MFCD00007935
White to off-white
1g
Powder
202.19
Videos
SDS