missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Nitrobenzeneboronic acid L-tartaric acid ester, 1M soln. in THF

Brand:  Alfa Aesar™ H55216.AA

Code : NEW

Additional Details : CAS Number : 467443-01-2 Weight : 0.00005kg Transport : UN number : 2056 Chem class : 3 Pack group : II

Quantity 5mL

 View more versions of this product

Product Code. 15471976

  • £49.60 / 5mL


Estimated Shipment date
from Supplier 18-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Nitrobenzeneboronic acid L-tartaric acid ester
280.99
UN2056
467443-01-2
MFCD09750458
3-Nitrophenylboronic acid L-tartaric acid ester, 1M soln. in THF

Specifications

3-Nitrobenzeneboronic acid L-tartaric acid ester
1.053g/mL
Colorless to brown
467443-01-2
1.4508
MFCD09750458
3-Nitrophenylboronic acid L-tartaric acid ester, 1M soln. in THF
1M soln. in THF
Moisture Sensitive
5mL
Liquid
280.99
UN2056
Videos
SDS