missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-5-(trifluoromethyl)pyridin-2-ol, 97%, Thermo Scientific™

Catalog Number p-2700245
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
33252-64-1
C6H3F3N2O3
208.096
MFCD00153193
JYXKHKBZLLIWEV-UHFFFAOYSA-N
2775093
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
This item is not returnable. View return policy
33252-64-1
C6H3F3N2O3
208.096
MFCD00153193
JYXKHKBZLLIWEV-UHFFFAOYSA-N
2775093
3-nitro-5-(trifluoromethyl)-1H-pyridin-2-one
C1=C(C(=O)NC=C1C(F)(F)F)[N+](=O)[O-]
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-hydroxy-3-nitro-5-trifluoromethyl pyridine, 3-nitro-5-trifluoromethyl pyridin-2 1h-one, 3-nitro-5-trifluoromethyl pyridin-2-ol, 3-nitro-5-trifluoromethyl-2-pyridinone, 2 1h-pyridinone,3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitropyridin-2-ol, 3-nitro-5-trifluoromethyl-1h-pyridin-2-one, 2 1h-pyridinone, 3-nitro-5-trifluoromethyl, 3-nitro-5-trifluoromethyl hydropyridin-2-one, pubchem9284
Formula Weight 208.09
Percent Purity 97%
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)pyridin-2-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'