missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-n-Hexylthiophene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
Solubility
Partly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.
Solubility
Partly soluble in water.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications
Density | 0.936 |
Boiling Point | 65°C (0.45 mmHg) |
Flash Point | 37°C (99°F) |
Odor | Stench |
Refractive Index | 1.496 |
Quantity | 1 g |
UN Number | UN1993 |
Beilstein | 1617129 |
Solubility Information | Partly soluble in water. |
Formula Weight | 168.3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'