missing translation for 'onlineSavingsMsg'
Learn More

3-n-Hexylthiophene, 98%, Thermo Scientific Chemicals

Catalog Number 15402288
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1693-86-3
C10H16S
168.298
MFCD00143181
JEDHEMYZURJGRQ-UHFFFAOYSA-N
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
566849
3-hexylthiophene
CCCCCCC1=CSC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1693-86-3
C10H16S
168.298
MFCD00143181
JEDHEMYZURJGRQ-UHFFFAOYSA-N
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
566849
3-hexylthiophene
CCCCCCC1=CSC=C1

3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-n-Hexylthiophene is a conducting polymer precursor. may be used in the synthesis of poly(3-hexylthiophene). MgKR X-ray and low energy electron induced oligomerization of physisorbed layers of 3-hexylthiophene condensed on clean gold results in the formation of an organic film.

Solubility
Partly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.936
Boiling Point 65°C (0.45 mmHg)
Flash Point 37°C (99°F)
Odor Stench
Refractive Index 1.496
Quantity 5 g
UN Number UN1993
Beilstein 1617129
Solubility Information Partly soluble in water.
Formula Weight 168.3
Percent Purity ≥99%
Chemical Name or Material 3-n-Hexylthiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'