missing translation for 'onlineSavingsMsg'
Learn More

3-n-Decylthiophene, 97%, Thermo Scientific™

Catalog Number 15401438
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
65016-55-9
C14H24S
224.406
MFCD00143180
JAYBIBLZTQMCAY-UHFFFAOYSA-N
3-n-decylthiophene, 3-decylthiophene, thiophene, 3-decyl, acmc-1bell, ksc491i4d
566842
3-decylthiophene
CCCCCCCCCCC1=CSC=C1
This item is not returnable. View return policy
65016-55-9
C14H24S
224.406
MFCD00143180
JAYBIBLZTQMCAY-UHFFFAOYSA-N
3-n-decylthiophene, 3-decylthiophene, thiophene, 3-decyl, acmc-1bell, ksc491i4d
566842
3-decylthiophene
CCCCCCCCCCC1=CSC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.912
Boiling Point 119°C to 120°C (1.2 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.489
Quantity 25 g
Formula Weight 224.4
Percent Purity 97%
Chemical Name or Material 3-n-Decylthiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'