missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-n-Butyl-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63046.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459498

  • £427.18 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-n-Butyl-1-methyl-2-thiohydantoin
186.27
MFCD27665228
3-n-Butyl-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-n-Butyl-1-methyl-2-thiohydantoin
95%
MFCD27665228
3-n-Butyl-1-methyl-2-thioxo-4-imidazolidinone
Solid
186.27
5g
Videos
SDS