missing translation for 'onlineSavingsMsg'
Learn More

3-MPPI, Tocris Bioscience™

α1 ligand

Brand:  Tocris (Bio-Techne) 058110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13427783

  • £160.23 / 10mg

Estimated Shipment date
from Supplier 06-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Very potent ligand for α1 sites (Ki for displacement of prazosin = 0.2 nM). Displays different binding properties for each α1 subtype (pKi values are 8.74, 9.44 and 9.57 for α1B, α1D and α1A adrenoceptors respectively). Also binds to 5-HT1A sites (Ki = 50 nM).
Specifications

Chemical Identifiers

133399-65-2
419.485
3-mppi, tocris-0581, biomol-nt_000127, 3-2-4-2-methoxyphenyl piperazino ethyl-1h-pyrimido 5,4-b indole-2,4 3h,5h-dione, 2-2-4-2-methoxy-phenyl-piperazin-1-yl-ethyl-4,9-dihydro-2,4,9-triaza-fluorene-1,3-dione, 3-2-4-2-methoxyphenyl piperazin-1-yl ethyl-1h-pyrimido 5,4-b indole-2,4 3h,5h-dione, 3-2-4-2-methoxyphenyl piperazin-1-yl ethyl-1,5-dihydropyrimido 5,4-b indole-2,4-dione, 3-2-4-2-methoxy-phenyl-piperazin-1-yl-ethyl-1,5-dihydro-pyrimido 5,4-b indole-2,4-dione
3-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-1,5-dihydropyrimido[5,4-b]indole-2,4-dione
C23H25N5O3
AQASGOHUMGAWJJ-UHFFFAOYSA-N
4013695
COC1=CC=CC=C1N2CCN(CC2)CCN3C(=O)C4=C(C5=CC=CC=C5N4)NC3=O

Specifications

10mg
133399-65-2
3-mppi, tocris-0581, biomol-nt_000127, 3-2-4-2-methoxyphenyl piperazino ethyl-1h-pyrimido 5,4-b indole-2,4 3h,5h-dione, 2-2-4-2-methoxy-phenyl-piperazin-1-yl-ethyl-4,9-dihydro-2,4,9-triaza-fluorene-1,3-dione, 3-2-4-2-methoxyphenyl piperazin-1-yl ethyl-1h-pyrimido 5,4-b indole-2,4 3h,5h-dione, 3-2-4-2-methoxyphenyl piperazin-1-yl ethyl-1,5-dihydropyrimido 5,4-b indole-2,4-dione, 3-2-4-2-methoxy-phenyl-piperazin-1-yl-ethyl-1,5-dihydro-pyrimido 5,4-b indole-2,4-dione
COC1=CC=CC=C1N2CCN(CC2)CCN3C(=O)C4=C(C5=CC=CC=C5N4)NC3=O
419.485
419.48
3-MPPI
C23H25N5O3
AQASGOHUMGAWJJ-UHFFFAOYSA-N
3-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-1,5-dihydropyrimido[5,4-b]indole-2,4-dione
4013695
>98%
Videos
Documents