missing translation for 'onlineSavingsMsg'
Learn More

(3-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Catalog Number p-3109876
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
145127-38-4
C11H15NO2
193.25
MFCD07772812
MXBVALXTJZMIJB-UHFFFAOYSA-N
7162074
(3-morpholin-4-ylphenyl)methanol
OCC1=CC(=CC=C1)N1CCOCC1
This item is not returnable. View return policy
145127-38-4
C11H15NO2
193.25
MFCD07772812
MXBVALXTJZMIJB-UHFFFAOYSA-N
7162074
(3-morpholin-4-ylphenyl)methanol
OCC1=CC(=CC=C1)N1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-morpholinophenyl methanol, 3-morpholinobenzyl alcohol, 3-morpholin-4-yl phenyl methanol, 3-morpholin-4-ylphenyl methanol, benzenemethanol,3-4-morpholinyl, benzenemethanol, 3-4-morpholinyl, 3-morpholinobenzylalcohol, acmc-20n4f1, 3-morpholin-4-ylphenyl methan-1-ol
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (3-Morpholinophenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'