missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Morpholinophenol, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
IUPAC Name | 3-morpholin-4-ylphenol |
Formula Weight | 179.22 |
Percent Purity | 97% |
Chemical Name or Material | 3-Morpholinophenol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'