missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholinomethyl)benzaldehyde, 95%, Thermo Scientific™

Catalog Number p-4510611
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
446866-83-7
C12H15NO2
205.257
KMYNYFKOOXFQGB-UHFFFAOYSA-N
3-morpholinomethyl benzaldehyde, 3-morpholin-4-ylmethyl benzaldehyde, benzaldehyde,3-4-morpholinylmethyl, 3-morpholin-4-ylmethyl-benzaldehyde, 3-4-morpholinylmethyl benzaldehyde, 3-morpholinomethylbenzaldehyde, 3-morpholin-4-yl methyl benzaldehyde
7164603
3-(morpholin-4-ylmethyl)benzaldehyde
C1COCCN1CC2=CC=CC(=C2)C=O
This item is not returnable. View return policy
446866-83-7
C12H15NO2
205.257
KMYNYFKOOXFQGB-UHFFFAOYSA-N
3-morpholinomethyl benzaldehyde, 3-morpholin-4-ylmethyl benzaldehyde, benzaldehyde,3-4-morpholinylmethyl, 3-morpholin-4-ylmethyl-benzaldehyde, 3-4-morpholinylmethyl benzaldehyde, 3-morpholinomethylbenzaldehyde, 3-morpholin-4-yl methyl benzaldehyde
7164603
3-(morpholin-4-ylmethyl)benzaldehyde
C1COCCN1CC2=CC=CC(=C2)C=O
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 205.26
Percent Purity 95%
Chemical Name or Material 3-(Morpholinomethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'