missing translation for 'onlineSavingsMsg'
Learn More

3-(Morpholin-4-ylmethyl)aniline, 95%, Thermo Scientific™

Catalog Number p-4510615
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
123207-48-7
C11H16N2O
192.262
IHHKTIKNEDOMIK-UHFFFAOYSA-N
3-morpholinomethyl aniline, 3-morpholin-4-ylmethyl aniline, 3-morpholin-4-ylmethyl-phenylamine, benzenamine,3-4-morpholinylmethyl, benzenamine, 3-4-morpholinylmethyl, 3-morpholin-4-yl methyl aniline, 3-4-morpholinylmethyl aniline, 3-morpholin-4-ylmethyl-aniline, 3-morpholin-4-ylmethyl phenylamine, 3-morpholinomethyl-aniline
1096406
3-(morpholin-4-ylmethyl)aniline
C1COCCN1CC2=CC(=CC=C2)N
This item is not returnable. View return policy
123207-48-7
C11H16N2O
192.262
IHHKTIKNEDOMIK-UHFFFAOYSA-N
3-morpholinomethyl aniline, 3-morpholin-4-ylmethyl aniline, 3-morpholin-4-ylmethyl-phenylamine, benzenamine,3-4-morpholinylmethyl, benzenamine, 3-4-morpholinylmethyl, 3-morpholin-4-yl methyl aniline, 3-4-morpholinylmethyl aniline, 3-morpholin-4-ylmethyl-aniline, 3-morpholin-4-ylmethyl phenylamine, 3-morpholinomethyl-aniline
1096406
3-(morpholin-4-ylmethyl)aniline
C1COCCN1CC2=CC(=CC=C2)N
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 3-(Morpholin-4-ylmethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'