missing translation for 'onlineSavingsMsg'
Learn More

3-Methylxanthine, 97%, ACROS Organics™

Brand:  Acros Organics 206930010

Code : NEW

Additional Details : CAS Number : 1076-22-8 Weight : 0.01000kg


Quantity 1g

 View more versions of this product

Product Code. 13438458

  • £127.00 / 1g

Estimated Shipment
09-02-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1076-22-8
166.14
3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
CHEBI:62207
CN1C2=C(C(=O)NC1=O)NC=N2
C6H6N4O2
GMSNIKWWOQHZGF-UHFFFAOYSA-N
70639
3-methyl-7H-purine-2,6-dione

Specifications

3-Methylxanthine
>300.0°C
1076-22-8
C6H6N4O2
3-methylxanthine, 3-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-3-methylpurine, xanthine, 3-methyl, 3 mx, 3-methyl xanthine, 1h-purine-2,6-dione, 3,7-dihydro-3-methyl, unii-ws6x982oec, 3-methyl-2,3,6,7-tetrahydro-1h-purine-2,6-dione, ccris 5817
GMSNIKWWOQHZGF-UHFFFAOYSA-N
3-methyl-7H-purine-2,6-dione
70639
166.14
Glass bottle
1g
96% min. (HPLC)
26,453
Solubility in water: insoluble
CN1C2=C(C(=O)NC1=O)NC=N2
166.14
CHEBI:62207
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Methylxanthine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents