missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)hexanol, 99%, Thermo Scientific™

Catalog Number p-7026016
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
51755-66-9
C7H16OS
148.26
MFCD00010674
JSASXSHMJYRPCM-UHFFFAOYNA-N
3-methylthio-1-hexanol, 3-methylthio hexanol, 1-hexanol, 3-methylthio, 3-methylthio hexan-1-ol, 3-methylthiohexanol, 3-methylsulfanyl hexan-1-ol, r-3-methylthio-1-hexanol, 3-methylmercapto-1-hexanol, fema no. 3438
65413
CCCC(CCO)SC
This item is not returnable. View return policy
51755-66-9
C7H16OS
148.26
MFCD00010674
JSASXSHMJYRPCM-UHFFFAOYNA-N
3-methylthio-1-hexanol, 3-methylthio hexanol, 1-hexanol, 3-methylthio, 3-methylthio hexan-1-ol, 3-methylthiohexanol, 3-methylsulfanyl hexan-1-ol, r-3-methylthio-1-hexanol, 3-methylmercapto-1-hexanol, fema no. 3438
65413
CCCC(CCO)SC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.966
Boiling Point 61°C to 62°C (10 mmHg)
Flash Point 107°C (226°F)
Linear Formula CH3CH2CH2CH(SCH3)CH2CH2OH
Refractive Index 1.4759
Quantity 5 g
UN Number UN3334
IUPAC Name 3-methylsulfanylhexan-1-ol
Formula Weight 148.26
Percent Purity 99%
Chemical Name or Material 3-(Methylthio)hexanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'