missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)aniline, 97%, ACROS Organics™

Catalog Number p-165036
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1783-81-9
C7H9NS
139.216
MFCD00007793
KCHLDNLIJVSRPK-UHFFFAOYSA-N
3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
15693
3-methylsulfanylaniline
CSC1=CC=CC(=C1)N
This item is not returnable. View return policy
1783-81-9
C7H9NS
139.216
MFCD00007793
KCHLDNLIJVSRPK-UHFFFAOYSA-N
3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
15693
3-methylsulfanylaniline
CSC1=CC=CC(=C1)N
CAS Min % 96.0
CAS Max % 100.0
Color Brown
Density 1.1300g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3SC6H4NH2
Refractive Index 1.6410 to 1.6450 (20°C, 589nm)
Quantity 10 g
Beilstein 13, IV, 1007
Formula Weight 139.21
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)aniline, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'