missing translation for 'onlineSavingsMsg'
Learn More

3-Methylsalicylic acid, 99%, ACROS Organics™

Brand:  Acros Organics 173341000

Code : 29

Additional Details : CAS Number : 83-40-9 Weight : 0.15000kg

Quantity 100g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10588863

  • £11.80 / 100g


Estimated Shipment
14-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Methylsalicylic acid
98.5
99%
C8H8O3
152.15
83-40-9
100.0
99%
CH3C6H3(OH)CO2H
MFCD00002448

Specifications

3-Methylsalicylic acid
83-40-9
100.0
Plastic bottle
Fine Powder
99%
CH3C6H3(OH)CO2H
152.15
10, 220
164.0° to 167.0°C
2-Hydroxy-3-methylbenzoic acid
100g
98.5
Authentic
Solubility in water: 1.5g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in chloroform, alcohol and ether
99%
C8H8O3
0.3% max. (K.F.)
MFCD00002448
15, 2570
Almost white
Videos
SDS