missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpicolinic acid, 98%, Thermo Scientific™

Catalog Number p-4089680
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4021-07-2
C7H7NO2
137.14
MFCD00191553
LMHIBYREWJHKNZ-UHFFFAOYSA-N
3-methylpyridine-2-carboxylic acid
This item is not returnable. View return policy
4021-07-2
C7H7NO2
137.14
MFCD00191553
LMHIBYREWJHKNZ-UHFFFAOYSA-N
3-methylpyridine-2-carboxylic acid
CAS Min % 97.5
CAS Max % 100.0
Melting Point 118.5°C to 121.5°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Synonym 3-methylpicolinic acid, 3-methyl-2-pyridinecarboxylic acid, 3-methyl-2-picolinic acid, 2-pyridinecarboxylic acid, 3-methyl, 3-methyl-a-picolinic acid, 3-methyl-alpha-picolinic acid, 3-methylpyridine-2-carboxylicacid, methylpyridincarbonsaure, pubchem14396, pubchem17659
SMILES CC1=C(N=CC=C1)C(=O)O
PubChem CID 268757
Formula Weight 137.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylpicolinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylpicolinic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'