Learn More
Thermo Scientific Maybridge 3-Methylisoxazole-5-carbaldehyde, 1g, 95%
Brand: Thermo Scientific Maybridge CC27004DA
3-Methylisoxazole-5-carbaldehyde, 1g, 95%
Code : 31
Additional Details : CAS Number : 70753-36-5 Weight : 0.05100kg
Brand: Thermo Scientific Maybridge CC27004DA
3-Methylisoxazole-5-carbaldehyde, 1g, 95%
Code : 31
Additional Details : CAS Number : 70753-36-5 Weight : 0.05100kg