missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylisoxazole-5-carbaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-methylisoxazole-5-carbaldehyde, 5-isoxazolecarboxaldehyde, 3-methyl, 3-methyl-5-isoxazolecarbaldehyde, 3-methyl-5-isoxazolecarboxaldehyde, 5-isoxazolecarboxaldehyde,3-methyl, 3-methyl-5-formyl-isoxazole, 3-methylisoxazol-5-carbaldehyde, 3-methyl-isoxazole-5-carbaldehyde, 3-methylisoxazole-5-carboxaldehyde, 5-isoxazolecarboxaldehyde, 3-methyl-6ci, 7ci, 9ci |
Formula Weight | 111.1 |
Percent Purity | 95% |
Chemical Name or Material | 3-Methylisoxazole-5-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'