missing translation for 'onlineSavingsMsg'
Learn More

3-Methylglutaric anhydride, 97%, Thermo Scientific Chemicals

Catalog Number p-163076
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4166-53-4
C6H8O3
128.13
MFCD00006683
MGICRVTUCPFQQZ-UHFFFAOYSA-N
3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
77828
4-methyloxane-2,6-dione
CC1CC(=O)OC(=O)C1
This item is not returnable. View return policy
4166-53-4
C6H8O3
128.13
MFCD00006683
MGICRVTUCPFQQZ-UHFFFAOYSA-N
3-methylglutaric anhydride, 4-methyldihydro-2h-pyran-2,6 3h-dione, 3-methylglutaricanhydride, .beta.-methylglutaric anhydride, acmc-1ago6, 2h-pyran-2,6 3h-dione, dihydro-4-methyl, 3-methyl glutaric anhydride, beta-methylglutaric anhydride, 4-methyltetrahydropyran-2,6-dione
77828
4-methyloxane-2,6-dione
CC1CC(=O)OC(=O)C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color White
Boiling Point 180°C to 182°C (25.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 415
Solubility Information Solubility in water: may decompose
Formula Weight 128.13
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 3-Methylglutaric anhydride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methylglutaric anhydride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'